Learn More
Invitrogen™ WDR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588389
Description
Immunogen sequence: YINYLDRNNP SKPLHVIKGH SKSIQCLTVH KNGGKSYIYS GSHDGHINYW DSETGENDSF AGKGHTNQVS RMTVDESGQL ISCSMDDTVR YTSLMLRDYS GQGVVKLDVQ PKCVAVGPGG YAVVVCIGQI VLLKDQRKCF SIDNPGYEPE VVAVHPGGDT VAIGGVDGNV RLYSILGTTL KDEGKLLEAK GPVTDVAYSH D; Positive Samples: Raji, HT-29, U-87MG, MCF7, NIH/3T3, Mouse brain, Mouse liver, Rat heart, Rat spinal cord; Cellular Location: Cell projection, Cytoplasm, cytoskeleton, podosome.
This gene encodes a protein containing 9 WD repeats. WD repeats are approximately 30- to 40-amino acid domains containing several conserved residues, mostly including a trp-asp at the C-terminal end. WD domains are involved in protein-protein interactions. The encoded protein may help induce the disassembly of actin filaments. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| WDR1 | |
| Polyclonal | |
| Unconjugated | |
| WDR1 | |
| Actin-interacting protein 1; Aip1; D5Wsu185e; epididymis secretory protein Li 52; HEL-S-52; NORI-1; rede; WD repeat domain 1; WD repeat protein 1; WD repeat-containing protein 1; WD40 repeat protein 1; Wdr1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 22388, 360950, 9948 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 11.5 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O75083, O88342, Q5RKI0 | |
| WDR1 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.