missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WDPCP Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560627
This item is not returnable.
View return policy
Description
Immunogen sequence: DIIFQYIHCD EIYEAINILS SMNWDTLGHQ CFISMSAIVN HLLRQKLTPE REAQLETSLG TFYAPTRPLL DSTILEYRDQ ISKYARRF Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 80%.
The C2ORF86 gene encodes an ortholog of Drosophila 'fritz,' a cytoplasmic WD40 repeat protein that controls planar cell polarity.
Specifications
| WDPCP | |
| Polyclonal | |
| Unconjugated | |
| WDPCP | |
| AV249152; Bardet-Biedl syndrome 15 protein; BBS15; C2orf86; CHDTHP; FRITZ; FRTZ; hFRTZ; homoloc-13; homolog-13; mFrtz; WD repeat containing planar cell polarity effector; WD repeat-containing and planar cell polarity effector protein; WD repeat-containing and planar cell polarity effector protein fritz homolog; WD repeat-containing protein C2orf86; WD repeat-containing protein C2orf86 homolog; Wdpcp | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51057 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O95876 | |
| WDPCP | |
| Recombinant protein corresponding to Human WDPCP. Recombinant protein control fragment (Product #RP-99655). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction