missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDFY3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55009-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody is specific to WDFY3. The 50 kDa band in WB is an isoform of WDFY3.
The ubiquitously expressed cellular upstream stimulatory factor (USF) consists of 43kDa (USF-1) and 44kDa (USF-2) polypeptides which independently exhibit site-specific DNA binding and are members of the c-Myc-related family of regulatory factors containing helix-loop-helix domains. USF also contains a leucine repeat that is required for efficient DNA binding.Specifications
| WDFY3 | |
| Polyclonal | |
| Unconjugated | |
| WDFY3 | |
| ALFY, Autophagy linked FYVE protein, KIAA0993, WDFY3, ZFYVE25 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23001, 72145 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6VNB8, Q8IZQ1 | |
| Wdfy3 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction