missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WDFY3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568443
This item is not returnable.
View return policy
Description
WDFY3 Polyclonal Antibody for Western Blot, IHC (P)
The ubiquitously expressed cellular upstream stimulatory factor (USF) consists of 43kDa (USF-1) and 44kDa (USF-2) polypeptides which independently exhibit site-specific DNA binding and are members of the c-Myc-related family of regulatory factors containing helix-loop-helix domains. USF also contains a leucine repeat that is required for efficient DNA binding.
Specifications
| WDFY3 | |
| Polyclonal | |
| Unconjugated | |
| WDFY3 | |
| 2610509D04Rik; ALFY; Autophagy-linked FYVE protein; AW319683; B930017C24; Bchs; beach domain, WD repeat and FYVE domain-containing protein 1; BWF1; D5Ertd66e; galactosyltransferase 3 beta 1, 4; Ggtb3; KIAA0993; LOW QUALITY PROTEIN: WD repeat and FYVE domain-containing protein 3; mKIAA0993; WD repeat and FYVE domain containing 3; WD repeat and FYVE domain-containing protein 3; WDFY3; ZFYVE25 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23001 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1.6 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q8IZQ1 | |
| WDFY3 | |
| Synthetic protein corresponding to residues near the C terminal of human WDFY3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction