missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WDFY3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA577795
This item is not returnable.
View return policy
Description
A 1:1,000 dilution of PA5-77795 was sufficient for detection of WDFY3 in 15 μg of Human HeLa Cell lysates by ECL immunoblot analysis using goat anti-rabbit IgG:HRP as the secondary antibody. Detects approximately 400 kDa.
The ubiquitously expressed cellular upstream stimulatory factor (USF) consists of 43kDa (USF-1) and 44kDa (USF-2) polypeptides which independently exhibit site-specific DNA binding and are members of the c-Myc-related family of regulatory factors containing helix-loop-helix domains. USF also contains a leucine repeat that is required for efficient DNA binding.
Specifications
| WDFY3 | |
| Polyclonal | |
| Unconjugated | |
| WDFY3 | |
| 2610509D04Rik; ALFY; Autophagy-linked FYVE protein; AW319683; B930017C24; Bchs; beach domain, WD repeat and FYVE domain-containing protein 1; BWF1; D5Ertd66e; galactosyltransferase 3 beta 1, 4; Ggtb3; KIAA0993; LOW QUALITY PROTEIN: WD repeat and FYVE domain-containing protein 3; mKIAA0993; WD repeat and FYVE domain containing 3; WD repeat and FYVE domain-containing protein 3; WDFY3; ZFYVE25 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23001, 305164 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.4 | |
| Q8IZQ1 | |
| WDFY3 | |
| Synthetic peptide from the N-terminal of Human WDFY3 (aa 533-544). | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction