missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WDFY1/2 Recombinant Rabbit Monoclonal Antibody (HL1989)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556919
This item is not returnable.
View return policy
Description
WDFY1/2 Recombinant Monoclonal Antibody for Western Blot
WD-repeats are motifs that are found in a variety of proteins and are characterized by a conserved core of 40-60 amino acids that commonly form a tertiary propeller structure. While proteins that contain WD-repeats participate in a wide range of cellular functions, they are generally involved in regulatory mechanisms concerning chromatin assembly, cell cycle control, signal transduction, RNA processing, apoptosis and vesicular trafficking. WDFY1 positively regulates TLR3- and TLR4-mediated signaling pathways by bridging the interaction between TLR3 or TLR4 and TICAM1. WDFY1 Promotes TLR3/4 ligand-induced activation of transcription factors IRF3 and NF-kappa-B, as well as the production of IFN-beta and inflammatory cytokines. WDFY2 acts in an adapter protein-like fashion to mediate the interaction between the kinase PRKCZ and its substrate VAMP2 and increases the PRKCZ-dependent phosphorylation of VAMP2. WDFY2 positively regulates adipocyte differentiation, by facilitating the phosphorylation and thus inactivation of the anti-adipogenetic transcription factor FOXO1 by the kinase AKT1.
Specifications
| WDFY1/2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q8IWB7 | |
| WDFY1 | |
| Recombinant fragment of human WDFY1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| HL1989 | |
| Unconjugated | |
| WDFY1 | |
| 1700013B03Rik; 1700120F24Rik; FENS1; FENS-1; FYVE domain-containing protein localized to endosomes 1; Jr1; KIAA1435; mKIAA1435; Phosphoinositide-binding protein 1; phosphoinositide-binding protein SR1; WD repeat and FYVE domain containing 1; WD repeat and FYVE domain-containing protein 1; WD40 and FYVE domain containing 1; WD40- and FYVE domain-containing protein 1; WDF1; WDFY1; ZFYVE17; Zinc finger FYVE domain-containing protein 17 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 57590 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction