missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WC1 Monoclonal Antibody (CC101)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA528515
This item is not returnable.
View return policy
Description
Mouse anti Bovine WC1 antibody, clone CC101, recognizes a subset of WC1+ T-cells expressing the WC1.1 isoform (MacHugh et al. 1993).
WC1 is approximately 215/300 kDa antigen expressed on the majority of gamma/delta T lymphocytes. These cells also express low levels of CD5, but are negative for other B and T cell markers. The WC1 antigen is reported to be involved in a number of processes, including activation of gamma/delta T cells and the development of a Th1-biased acquired immune response.
Specifications
| WC1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| P30205 | |
| CD163L1 | |
| Con A stimulated bovine lymphocytes. | |
| 100 μg | |
| Primary | |
| Bovine | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation | |
| CC101 | |
| Unconjugated | |
| CD163L1 | |
| antigen WC1.1; BoWC1.1; CD163 molecule-like 1; CD163L1; scavenger receptor cysteine-rich type 1 protein M160; scavenger-receptor protein; WC1; WC1 tail; WC1.1; WC1.1 molecule; WC1.1 tail; WC1-2; WC1-5; WC1-5 transmembrane protein | |
| Mouse | |
| Protein G | |
| RUO | |
| 338056 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction