missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WC1(+) gamma delta T cell Antibody (BAQ159A), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61085
This item is not returnable.
View return policy
Description
WC1(+) gamma delta T cell Monoclonal specifically detects WC1(+) gamma delta T cell in Bovine, Water Buffalo samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| WC1(+) gamma delta T cell | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry 1:10 - 1:1000 | |
| Mouse | |
| Unpurified | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Flow Cytometry, Immunohistochemistry, Functional Assay | |
| BAQ159A | |
| Unconjugated | |
| Ascites with 0.09% Sodium Azide | |
| Bovine ConA stimulated mononuclear leukocytes | |
| 0.1 mg | |
| Bovine, Water Buffalo | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction