missing translation for 'onlineSavingsMsg'
Learn More

WBSCR22 Antibody (2E12), Novus Biologicals™

Product Code. 18414709 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18414709 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18414709 Supplier Novus Biologicals Supplier No. H00114049M11

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

WBSCR22 Monoclonal antibody specifically detects WBSCR22 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen WBSCR22
Applications Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Sandwich ELISA
Classification Monoclonal
Clone 2E12
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_059998
Gene Alias EC 2.1.1.-, FLJ44236, HASJ4442, HUSSY-3, MGC19709, MGC2022, MGC5140, PP3381, WBMT, Williams Beuren syndrome chromosome region 22, Williams Beuren syndrome chromosome region 22 protein, Williams-Beuren candidate region putative methyltransferase, Williams-Beuren syndrome chromosomal region 22 protein
Host Species Mouse
Immunogen WBSCR22 (NP_059998.2, 1 a.a. ∽ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MASRGRRPEHGGPPELFYDETEARKYVRNSRMIDIQTRMAGRALELLYLPENKPCYLLDIGCGTGLSGSYLSDEGHYWVGLDISPAMLDEAVDREIEGDLLLGDMGQGIP
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114049
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.