missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WBSCR17 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12659-50ul
This item is not returnable.
View return policy
Description
WBSCR17 Polyclonal antibody specifically detects WBSCR17 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| WBSCR17 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| DKFZp434I2216, EC 2.4.1.41, GalNAc-T-like protein 3, GALNT16, GALNT20, GALNTL3DKFZp761D2324, Polypeptide GalNAc transferase-like protein 3, polypeptide N-acetylgalactosaminyltransferase WBSCR17, pp-GalNAc-transferase 20, pp-GaNTase-like protein 3, Protein-UDP acetylgalactosaminyltransferase-like protein 3, putative polypeptide N-acetylgalactosaminyltransferase-like protein 3, UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase-like 3, UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase-like protein 3, UDP-N-acetyl-alpha-D-galactosamine:polypeptideN-acetylgalactosaminyltransferase 16, Williams-Beuren syndrome chromosomal region 17 protein, Williams-Beuren syndrome chromosome region 17 | |
| Produced in rabbits immunized with E. coli-derived Human WBSCR17 fragment | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 64409 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction