missing translation for 'onlineSavingsMsg'
Learn More

WBSCR16 Antibody, Novus Biologicals™

Product Code. 18409891 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18409891 25 μL 25µL
18119774 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18409891 Supplier Novus Biologicals Supplier No. NBP23183325ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

WBSCR16 Polyclonal specifically detects WBSCR16 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WBSCR16
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q96I51
Gene Alias DKFZp434D0421, MGC189739, MGC44931, RCC1-like G exchanging factor-like protein, Williams-Beuren syndrome chromosomal region 16 protein, Williams-Beuren syndrome chromosome region 16
Gene Symbols RCC1L
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: TLLSSKTADVTKVWGMGLNKDSQLGFHRSRKDKTRGYEYVLEPSPVSLPLDRPQETRVLQVSCGRAHSLVLTDREGVFSMGNNSYGQCGRKVVEN
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81554
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.