missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WBP4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558612
This item is not returnable.
View return policy
Description
Immunogen sequence: YCKCWIADNR PSVEFHERGK NHKENVAKRI SEIKQKSLDK AKEEEKASKE FAAMEAAALK AYQEDLKRLG LESEILEPSI TPVTSTIPPT STSNQQKEK Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 88%.
This gene encodes WW domain-containing binding protein 4. The WW domain represents a small and compact globular structure that interacts with proline-rich ligands. This encoded protein is a general spliceosomal protein that may play a role in cross-intron bridging of U1 and U2 snRNPs in the spliceosomal complex A.
Specifications
| WBP4 | |
| Polyclonal | |
| Unconjugated | |
| WBP4 | |
| AW545037; BB101031; domain-containing binding protein 4; FBP21; Fnbp21; formin binding protein 21; formin-binding protein 21; hypothetical protein LOC553723; MGC117310; Wbp4; WBP-4; WW domain binding protein 4; WW domain binding protein 4 (formin binding protein 21); WW domain-binding protein 4; WW domain-binding protein 4-like protein; WW domain-containing binding protein 4; WW domain-containing-binding protein 4; zgc:112384; zgc:112384 protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11193 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75554 | |
| WBP4 | |
| Recombinant protein corresponding to Human WBP4. Recombinant protein control fragment (Product #RP-97930). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction