missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WBP4 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18460G
This item is not returnable.
View return policy
Description
WBP4 Polyclonal antibody specifically detects WBP4 in Human samples. It is validated for Western Blot
Specifications
| WBP4 | |
| Polyclonal | |
| Western Blot | |
| domain-containing binding protein 4, WW domain binding protein 4 (formin binding protein 21) | |
| Recognizes region between residues Between Between 125 and 175.. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 11193 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction