missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WBP2NL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106387
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total WBP2NL.
May play a role in meotic resumption and pronuclear formation, mediated by a WW domain-signaling pathway during fertilization.
Specifications
| WBP2NL | |
| Polyclonal | |
| Unconjugated | |
| WBP2NL | |
| 4930521I23Rik; GRAMD7; PAWP; Postacrosomal sheath WW domain-binding protein; WBP2 N-terminal like; Wbp2nl; WW domain-binding protein 2-like | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 164684, 74716 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6ICG8, Q9D529 | |
| WBP2NL | |
| A synthesized peptide derived from human WBP2NL(Accession Q6ICG8), corresponding to amino acid residues G202-Y252. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction