missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WBP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111638
This item is not returnable.
View return policy
Description
Immunogen sequence: PYPGPMEPPV SGPDVPSTPA AEAKAAEAAA SAYYNPGNPH NVYMPTSQPP PPPYYP Highest antigen sequence indentity to the following orthologs: Rat - 93%, Mouse - 95%.
The globular WW domain is composed of 38 to 40 semiconserved amino acids shared by proteins of diverse functions including structural, regulatory, and signaling proteins. The domain is involved in mediating protein-protein interactions through the binding of polyproline ligands. WBP2 is a WW domain binding protein, which binds to the WW domain of Yes kinase-associated protein by its PY motifs. The function of this protein has not been determined.
Specifications
| WBP2 | |
| Polyclonal | |
| Unconjugated | |
| WBP2 | |
| GRAMD6; Wbp2; WBP-2; WW domain binding protein 2; WW domain-binding protein 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23558 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q969T9 | |
| WBP2 | |
| Recombinant Protein corresonding to Human WBP2. Recombinant protein control fragment (Product #RP-107655). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction