missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WBP11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10316-1-AP-150UL
This item is not returnable.
View return policy
Description
The predicted MW of this protein is 70 kDa, and this antibody specifically recognizes the 70 kDa protein.
This gene encodes a nuclear protein, which colocalizes with mRNA splicing factors and intermediate filament-containing perinuclear networks. This protein has 95% amino acid sequence identity to the mouse Wbp11 protein. It contains two proline-rich regions that bind to the WW domain of Npw38, a nuclear protein, and thus this protein is also called Npw38-binding protein NpwBP. The Npw38-NpwBP complex may function as a component of an mRNA factory in the nucleus.Specifications
| WBP11 | |
| Polyclonal | |
| Unconjugated | |
| WBP11 | |
| SIPP1, SNP70, WBP 11, WBP11, WW domain binding protein 11 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 297695, 51729, 60321 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.43 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5PQQ2, Q923D5, Q9Y2W2 | |
| Wbp11 | |
| WBP11 Fusion Protein Ag0366 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction