missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WBP11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531241
This item is not returnable.
View return policy
Description
Recommended positive controls: Jurkat, Raji. Predicted reactivity: Mouse (98%), Rat (99%), Xenopus laevis (85%), Rhesus Monkey (99%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Posttranslational modification of proteins by the addition of the small protein SUMO.
Specifications
| WBP11 | |
| Polyclonal | |
| Unconjugated | |
| WBP11 | |
| 2510026P17Rik; D6Wsu113e; DKFZp779M1063; hypothetical protein LOC402950; Npw38-binding protein; Npw38-binding protein NpwBP; NpwBP; PPP1R165; protein phosphatase 1, regulatory subunit 165; SH3 domain-binding protein; SH3 domain-binding protein SNP70; SIPP1; SNP70; Splicing factor that interacts with PQBP-1 and PP1; splicing factor, PQBP1 and PP1 interacting; Wbp11; WBP-11; WW domain binding protein 11; WW domain-binding protein 11; zgc:77390 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 297695, 51729, 60321 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.7 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q5PQQ2, Q923D5, Q9Y2W2 | |
| WBP11 | |
| Recombinant protein encompassing a sequence within the center region of human WBP11. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction