missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WBP11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97493-50ul
This item is not returnable.
View return policy
Description
WBP11 Polyclonal antibody specifically detects WBP11 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| WBP11 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| DKFZp779M1063, Npw38-binding protein, Npw38-binding protein NpwBP, NpwBP, NPWBPsplicing factor, PQBP1 and PP1 interacting, SH3 domain-binding protein SNP70, SIPP1WW domain-binding protein 11, SNP70, Splicing factor that interacts with PQBP-1 and PP1, WBP-11, WW domain binding protein 11 | |
| Produced in rabbits immunized with E. coli-derived Human WBP11 fragment. | |
| 50 μg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 51729 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction