Learn More
WBP1 Polyclonal Antibody
GREENER_CHOICE
WBP1 Polyclonal Antibody
Brand:  Invitrogen PA589083
Product Code. 16557111
  • £441.00 / 100µL
Description
WBP1 Polyclonal Antibody for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
WBP1
Polyclonal
Unconjugated
WBP1
AI255152; WBP1; WBP-1; WW domain binding protein 1; WW domain-binding protein 1
Rabbit
Affinity Chromatography
RUO
22377, 23559, 297381
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
0.93 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
P97764, Q96G27
WBP1
Recombinant fusion protein containing a sequence corresponding to amino acids 170-269 of human WBP1 (NP_0366091).
100 μL
Primary
Human, Mouse, Rat