Learn More
WBP1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11042-1-AP-150UL
 View more versions of this product
Product Code. 16877843
  • £349.00 / 150µL
Description
The globular WW domain, named for the conserved tryptophan residues in the protein motif present in various structural and regulatory proteins, is known to play a role in the mediation of protein-protein interactions. This gene encodes a ligand of the WW domain of the Yes kinase-associated protein. Readthrough transcription of the neighboring upstream gene, which encodes INO80 complex subunit B, into this gene generates a non-coding transcript.
Specifications
Specifications
WBP1
Polyclonal
Unconjugated
WBP1
WBP 1, WBP1, WW domain binding protein 1
Rabbit
Antigen Affinity Chromatography
RUO
23559
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q96G27
WBP1
WBP1 Fusion Protein Ag1478
150 μL