Learn More
WBP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA563396
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: DSGIELCPCP ASGEGEPVKE VRVSATLPDL EDYSPCALPP ESVPQIFPMG LSSSEG Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 73%.
The globular WW domain, named for the conserved tryptophan residues in the protein motif present in various structural and regulatory proteins, is known to play a role in the mediation of protein-protein interactions. This gene encodes a ligand of the WW domain of the Yes kinase-associated protein. Readthrough transcription of the neighboring upstream gene, which encodes INO80 complex subunit B, into this gene generates a non-coding transcript.Specifications
WBP1 | |
Polyclonal | |
Unconjugated | |
WBP1 | |
AI255152; WBP1; WBP-1; WW domain binding protein 1; WW domain-binding protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23559 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96G27 | |
WBP1 | |
Recombinant protein corresponding to Human WBP1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only