missing translation for 'onlineSavingsMsg'
Learn More

WAY-262611, MedChemExpress

Product Code. p-200099934
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16460697

Brand: Medchem Express HY11035

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress WAY-262611 is a wingless β-Catenin agonist that increases bone formation rate with an EC50 of 0.63 μM in TCF-Luciferase assay. WAY-262611 is also a Dkk1 inhibitor.

WAY-262611 is a wingless β-Catenin agonist that increases bone formation rate with an EC50 of 0.63 μM in TCF-Luciferase assay. WAY-262611 is also a Dkk1 inhibitor.

Specifications

Chemical Name or Material WAY-262611
CAS 1123231-07-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C20H22N4
Quantity 50 mg
Solubility Information DMSO : ≥ 42 mg/mL (131.90 mM)
SMILES NCC1CCN(C2=NC=CC(C3=CC=C4C=CC=CC4=C3)=N2)CC1
Molecular Weight (g/mol) 318.42
Formula Weight 318.42
Percent Purity 99.51%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.