missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ WAY 213613

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Potent, non-substrate inhibitor of EAAT2 (GLT-1) that displays > 44-fold selectivity over EAAT1 and EAAT3 (IC50 values are 85, 3787 and 5004 nM for EAAT2, EAAT3 and EAAT1 respectively). Exhibits no activity towards ionotropic and metabotropic glutamate receptors.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 415.19 |
| Purity | >99% |
| Molecular Weight (g/mol) | 415.191 |
| Chemical Name or Material | WAY 213613 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction