missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WAY-100635 Maleate Salt, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | N-[2-[4-(2-Methoxyphenyl)-1-piperazinyl]ethyl]-N-2-pyridinyl-Cyclohexanecarboxamide (2Z)-2-Butenedioate |
| CAS | 634908-75-1 |
| Molecular Formula | C25H34N4O2 . C4H4O4 |
| Quantity | 5 mg |
| Synonym | WAY-100635 Maleate Salt |
| InChI Formula | InChI=1S/C25H34N4O2.C4H4O4/c1-31-23-12-6-5-11-22(23)28-18-15-27(16-19-28)17-20-29(24-13-7-8-14-26-24)25(30)21-9-3-2-4-10-21;5-3(6)1-2-4(7)8/h5-8,11-14,21H,2-4,9-10,15-20H2,1H3;1-2H,(H,5,6)(H,7,8)/b;2-1- |
| SMILES | COc1ccccc1N2CCN(CCN(C(=O)C3CCCCC3)c4ccccn4)CC2.OC(=O)\C=C/C(=O)O |
| IUPAC Name | (Z)-but-2-enedioic acid;N-[2-[4-(2-methoxyphenyl)piperazin-1-yl]ethyl]-N-pyridin-2-ylcyclohexanecarboxamide |
| Molecular Weight (g/mol) | 422.6 |
| Formula Weight | 538.2791 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction