missing translation for 'onlineSavingsMsg'
Learn More

WAY-100635 Maleate, MedChemExpress

Product Code. p-200097919
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16484886

Brand: Medchem Express HY10349A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress WAY-100635 maleate is a potent and selective 5-hydroxytryptamine 1A (5-HT1A) receptor antagonist with an IC50 value of 0.91 nM and Ki value of 0.39 nM. WAY-100635 maleate has pIC50 values for 5-HT1A and α1-adrenergic receptors of 8.9 and 6.6, respectively. WAY-100635 maleate is also a potent dopamine D4 receptor agonist.

WAY-100635 maleate is a potent and selective 5-hydroxytryptamine 1A (5-HT1A) receptor antagonist with an IC50 value of 0.91 nM and Ki value of 0.39 nM. WAY-100635 maleate has pIC50 values for 5-HT1A and α1-adrenergic receptors of 8.9 and 6.6, respectively. WAY-100635 maleate is also a potent dopamine D4 receptor agonist.

Specifications

Chemical Name or Material WAY-100635 Maleate
CAS 1092679-51-0
Color White
Purity Grade Notes Research
Molecular Formula C29H38N4O6
Quantity 100 mg
Solubility Information DMSO : ≥ 250 mg/mL (464.13 mM) ∣H2O : 25 mg/mL (46.41 mM; Need ultrasonic)
SMILES O=C(O)/C=C\C(O)=O.O=C(N(C1=NC=CC=C1)CCN2CCN(CC2)C3=CC=CC=C3OC)C4CCCCC4.[1:1]
Molecular Weight (g/mol) 538.64
Formula Weight 538.64
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.