missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WAY-100635 Maleate, MedChemExpress
Description
WAY-100635 maleate is a potent and selective 5-hydroxytryptamine 1A (5-HT1A) receptor antagonist with an IC50 value of 0.91 nM and Ki value of 0.39 nM. WAY-100635 maleate has pIC50 values for 5-HT1A and α1-adrenergic receptors of 8.9 and 6.6, respectively. WAY-100635 maleate is also a potent dopamine D4 receptor agonist.
Specifications
Specifications
| Chemical Name or Material | WAY-100635 Maleate |
| CAS | 1092679-51-0 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C29H38N4O6 |
| Quantity | 100 mg |
| Solubility Information | DMSO : ≥ 250 mg/mL (464.13 mM) ∣H2O : 25 mg/mL (46.41 mM; Need ultrasonic) |
| SMILES | O=C(O)/C=C\C(O)=O.O=C(N(C1=NC=CC=C1)CCN2CCN(CC2)C3=CC=CC=C3OC)C4CCCCC4.[1:1] |
| Molecular Weight (g/mol) | 538.64 |
| Formula Weight | 538.64 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?