Learn More
Eppendorf™ Wax-Coated Freezer Boxes Dividers
Product Code. 13198333
missing translation for 'orderingAttributeHoverText'
Tube Array:
10 x 10
8 x 8
missing translation for 'unitSize'
Each
Product Code. 13198333
missing translation for 'mfr': Eppendorf™ D64
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Material Wax-coated
Shape Square
Type Box Divider
For Use With (Application) Ultra-Low Temperature Freezer Box
For Use With (Equipment) Freezer Cardboard Storage Boxes
Holds 64 Tubes
Quantity 1 Each
Tube Array 8 x 8