missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Portable pH Meter

Product Code. 11652420
Change view
Click to view available options
Range (pH):
-2 to 16
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range (pH) unitSize
11652420 -2 to 16 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11652420 Supplier Hanna Instruments™ Supplier No. HI8424

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Uses a BNC connector for the pH electrode input allowing for a wide selection of pH and ORP electrodes to be used. Hanna Instruments™ Portable pH Meter is compact, waterproof, and supplied complete with a protective carrying case, PEI plastic body pH electrode, stainless steel temperature probe, calibration buffer sachets, and cleaning solution sachets.

  • Automatic or manual temperature compensation
  • Automatic calibration using two buttons
  • Automatic buffer recognition at pH4.01, pH7.01 and pH10.01
  • With combination gel electrode and temperature probe
  • Waterproof Measure pH, mV (ISE and ORP ranges) and temperature.

Specifications

Accuracy (Temperature) ±0.4°C/±0.8°F
Accuracy (mV) ±0.2 to ±1
Accuracy (pH) ±0.01
For Use With (Application) Automatic calibration, Temperature Compensation, Buffer recognition
Range (pH) -2 to 16
Resolution (pH) 0.01
Range (mV) ±699.9 to ±1999
Range (ORP) ±699.9 to ±1999mV
Resolution (mV) 0.1 to 1mV
Calibration Points 4.01, 7.01, 10.01
Temperature Range (Metric) -20°C to +120°C
Resolution (Temperature) 0.1°C, 0.1°F
Temperature Probe Calibration HI 7662 stainless steel temperature probe with 1 m (3.3’) cable (included)
Display Type LCD
Power Supply Yes
Probe Type Cable
Resolution 0.01pH
Detection Range 0 to 14 pH
Temperature Compensation Automatic, –20 to 120°C
Battery Type 9V
Battery Life 150 hr.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.