missing translation for 'onlineSavingsMsg'
Learn More

Kolmi Hopen™ Waterproof, Polyethylene-coated Isolation Gown

Product Code. 17790139 Shop All Medicom Products
Click to view available options
:
Waterproof, Polyethylene-coated Isolation Gown; Closure Type: Tie; Color: Sky Blue; Disposable: Yes; Material: Polypropylene, Polyethylene; Size: X-Large
This item is not returnable. View return policy

Product Code. 17790139

Brand: Kolmi Hopen™ A69955

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Enjoy the comfort and solidity of this isolation gown without jeopardizing the high quality infection and fluid protection you need in the healthcare setting.

  • Large opening for passage of the head
  • Patented 'Flexneck' technology consisting of double elastic
  • Long sleeves with thumb holes that completely cover wrists
  • Waterproof, designed to protect healthcare professionals and patients from cross-infection

Specifications

Size X-Large
For Use With (Application) Medical
Gender Unisex
No. of Pockets None
Product Type General Purpose Gown
Material Polyethylene, Polypropylene
Color Sky Blue
Closure Type Back Ties
Disposable Single-use
Sterility Non-sterile
Autoclavable Not Autoclavable
No. per Pack 10 Pk.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.