Learn More
Labconco™ WaterPro™ Filters
Filters, prefilters and cartridges for Labconco WaterPro PS and RO Water Purification Systems.
Brand:  Labconco™ 9092700
Code : A0
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11844750
  • £81.50 / Each
Estimated Shipment: 15-07-2024
to see stock.
Add to basket
Description
Description
5-6 micron carbon prefilter.
Specifications
Specifications
Replacement prefilter
WaterPro™ mobile system (Labconco Cat. Nos. 90900-00, -01, 90901-00, -01 and 90902-00, -01)
WaterPro Filters