missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ WaterBugs™ Escherichia coli Strain C1

Product Code. 12986047
Change view
Click to view available options
Quantity:
10 Tests/Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12986047 10 Tests/Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
If you would like to order, a signed ATCC agreement is required.
Product Code. 12986047 Supplier Thermo Scientific™ Supplier No. R4751985
  • ONLINE EXCLUSIVE PRICE
    £536.00
    £481.93 / Pack of 1
    Save £54.07
    10% Off
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
If you would like to order, a signed ATCC agreement is required.

Thermo Scientific™ WaterBugs Escherichia coli Strain C1 is used for water/wastewater quality control testing; microorganisms are supplied in Quanti-Cult format.

Thermo Scientific™ WaterBugs Escherichia coli Strain C1 is used for water/wastewater quality control testing; microorganisms are supplied in Quanti-Cult format.

  • Deliver <50 CFUs in a single inoculum to mimic water samples
  • One test/reconstituted vial

Ordering Alerts:

Supplied with 10 preserved organism vials; 10 rehydration vials.

TRUSTED_SUSTAINABILITY

Specifications

Description Escherichia coli Strain C1
Format Quanti-Cult
Product Type Quality Control Organism
Target Organism Class Gram-negative Rods
Test Type Quantitative QC
Packaging Type 10 Test/Kit
Quantity 10 Tests/Kit

The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.