missing translation for 'onlineSavingsMsg'
Learn More

Water, Ultra Resi-analyzed, J.T. Baker™

Product Code. 15334575 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass
Unit Size:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15334575 4 L Glass 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15334575 Supplier Avantor J.T.Baker Supplier No. 421903

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 7732-18-5
Color Colorless
Melting Point 0°C
Boiling Point 100°C
Molecular Formula H2O
Synonym dihydrogen oxide, dihydrogen monoxide
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES O
IUPAC Name oxidane
Molecular Weight (g/mol) 18.015
PubChem CID 962
ChEBI CHEBI:15377
Formula Weight 18
Packaging Glass
Quantity 4 L
Physical Form Liquid
Chemical Name or Material Water
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.