missing translation for 'onlineSavingsMsg'
Learn More

Water, for HPLC

Product Code. p-216517 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Unit Size:
1 L
2.5 L
This item is not returnable. View return policy

Product Code. 10541344

Brand: Thermo Scientific Acros 389390010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7732-18-5 | H2O | 18.015 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by a SFI certified supplier.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Absorbance 0.004 max. at 254nm, 0.009 max. at 210nm, 0.05 max. at 200nm
Color Colorless
Boiling Point 100.0°C
Conductivity 1μS/cm Max.
Linear Formula H2O
Merck Index 15, 10237
Formula Weight 18.02
Grade HPLC
Packaging Glass Bottle
Residue after Evaporation 0.0003% max.
Viscosity 0.89 mPa.s (25°C)
Density 1.0000g/mL
Quantity 1 L
Refractive Index 1.3334
Specific Gravity 1
Physical Form Liquid
Chemical Name or Material Water
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.