Learn More
Palintest™ Water Testing
Product Code. 13159280
Click to view available options
Format:
Tablet
Unit Size:
Pack of 250
Product Code. 13159280
Brand: Palintest™ AS 096
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
These water testing tablets are ideal for quick and reliable testing in the field.
  • For calcium and magnesium ion results hardness expressed in ppm of calcium carbonate 
  • Ideal for quick and reliable testing in the field 
  • They are crushed in the water sample until a color change occurs and the number required allows results to be calculated in ppm or mg/L
  • Magnesium hardness is the difference between total and calcium hardness
  • Permanent hardness is the difference between total hardness and total alkalinity
  • In bottles of 250 tablets with instructions
Specifications
Format Tablet
Packaging Bottle
Quantity 250 Pack