Learn More
Palintest™ Water Testing Tablet
Product Code. p-8007361
Click to view available options
For Use With (Application):
Calcium Hardness
Chloride Ions
Total Alkalinity
Unit Size:
Pack of 250
Product Code. 13129280
Brand: Palintest™ AS 072
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Superior quality water testing tablet ideal for quick and reliable testing in the field.
  • Ideal for quick and reliable testing in the field
  • They are crushed in the water sample until a color change occurs and the number required allows results to be calculated in ppm or mg/L
  • Magnesium hardness is the difference between total and calcium hardness
  • Permanent hardness is the difference between total hardness and total alkalinity
  • In bottles of 250 tablets with instructions
Specifications
For Use With (Application) Total Alkalinity
Quantity 250 Pack
Type Water Testing Tablet