missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palintest™ Water Testing Tablet
Click to view available options
For Use With (Application):
Calcium Hardness
Chloride Ions
Total Alkalinity
Unit Size:
Pack of 250
Description
Ideal for quick and reliable testing in the field. They are crushed in the water sample until a color change occurs and the number required allows results to be calculated in ppm or mg/L. Magnesium hardness is the difference between total and calcium hardness. Permanent hardness is the difference between total hardness and total alkalinity. In bottles of 250 tablets with instructions.
Specifications
Specifications
| For Use With (Application) | Calcium Hardness |
| Quantity | 250 Pack |
| Type | Water Testing Tablet |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P363-Wash contaminated clothing before reuse.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction