Description
External, free-standing, set consisting of 20 L fresh water canister with pump and level sensor and 20 L waste water canister, for flexible and independent water supply.
Specifications
Specifications
| For Use With (Equipment) | KBF 130, KBF 260, KBF 470, KBF 720, KBF 1060, KBF 1600, KBF PRO 130, KBF PRO 260, KBF PRO 470, KBF PRO 720, KBF PRO 1060, KBF PRO 1600 |
| Material | Plastic |
| Type | Water Supply |
| Size | 20 L |