missing translation for 'onlineSavingsMsg'
Learn More

Water Supply QC Check Sample: Solids

Product Code. 30316797
Change view
Click to view available options
Quantity:
23 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30316797 23 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316797 Supplier VHG Labs Supplier No. VHGQWSSOL23
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Non-Filterable Residue∣Total Filterable Residue∣Total Solids
Analyte or Component Names Non-Filterable Residue∣Total Filterable Residue∣Total Solids
Shipping Condition Room Temperature
Solution Type Mixture
Solvent or Matrix Water
Industry Type Petrochemical Testing
Quantity 23 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.