missing translation for 'onlineSavingsMsg'
Learn More

Water Supply QC Check Sample: o-Phosphate Nutrients

Product Code. 30326733
Click to view available options
Quantity:
15 mL
Unit Size:
15mL
This item is not returnable. View return policy

Product Code. 30326733

Brand: VHG Labs VHGQWSONUT15

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ortho-phosphate
CAS 7558-79-4
Analyte or Component Names Ortho-phosphate
Shipping Condition Room Temperature
Molecular Formula 2 Na . H O4 P
Synonym Disodium phosphate anhydrous, Disodium hydrogen phosphate
InChI Formula InChI=1 S/2 Na.H3O4P/c;;1-5(2,3)4/h;;(H3,1,2,3,4)/q2*+1;/p-2
Solution Type Single Solution
SMILES [Na+].[Na+].OP(=O)([O-])[O-]
IUPAC Name disodium;hydrogen phosphate
Solvent or Matrix Dilute HCl
Industry Type Petrochemical Testing
Molecular Weight (g/mol) 141.96
Quantity 15 mL
Formula Weight 141.9408
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.