missing translation for 'onlineSavingsMsg'
Learn More

Water Supply QC Check Sample: Hardness

Product Code. 30321562
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30321562 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30321562 Supplier VHG Labs Supplier No. VHGQWSHRD250
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Calcium∣Calcium hardness∣Magnesium∣Sodium∣Total hardness
CAS 7440-70-2, 7439-95-4, 7440-23-5
Analyte or Component Names Calcium∣Calcium hardness∣Magnesium∣Sodium∣Total hardness
Shipping Condition Room Temperature
Solution Type Mixture
Element Multi-element
Solvent or Matrix Water
Industry Type Petrochemical Testing
Quantity 250 mL
Concentration or Composition (by Analyte or Components) Sodium 14 mg/L∣Magnesium 16.1 mg/L∣Calcium 65.4 mg/L∣Calcium hardness 163 mg/L∣Total hardness 230 mg/L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.