Learn More
BINDER Water Supply
Product Code. 30274306 Shop All BINDER Products
Change view
Click to view available options
For Use With (Equipment):
CBF with Active Humidification
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
30274306 CBF with Active Humidification Each
1 options
Product Code. 30274306
Brand: BINDER 80091192
Only null left
Add to basket
Add to basket
Set consisting of 6 bottles of sterile humidification water.
  • For CO2 incubators CB/CB-S
Specifications
For Use With (Equipment) CBF with Active Humidification
Material Plastic
Type Sterile Water