missing translation for 'onlineSavingsMsg'
Learn More

Water, for spectroscopy ACS, Thermo Scientific Chemicals

Product Code. p-4229433 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
This item is not returnable. View return policy

Product Code. 10371810

Brand: Thermo Scientific Acros 433382500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Synonym dihydrogen oxide, dihydrogen monoxide

Specifications

Absorbance 0.005 max. at 254nm, 0.005 max. at 300nm, 0.01 max. at 200nm, 0.01 max. at 210nm, Passes Test
Color Colorless
Boiling Point 100.0°C
Linear Formula H2O
Density 1.0000g/mL
Formula Weight 18.02
Grade ACS
Packaging Glass bottle
Refractive Index 1.3334
Specific Gravity 1
Viscosity 0.89 mPa.s (25°C)
Quantity 250 mL
Physical Form Liquid
Chemical Name or Material Water, For Spectroscopy
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.