Learn More
LABPLAS Water Sampling Kit Writing
Product Code. 17930115
Change view
Click to view available options
Volume (Metric):
100 mL
300 mL
500 mL
1000 mL
Unit Size:
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
17930115 300 mL Pack of 100
17910245 100 mL Pack of 100
17980225 500 mL Pack of 100
17980135 1000 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17930115 Supplier LABPLAS Supplier No. KWS33100
Only null left
Add to basket
Edge
Add to basket
Sterile water sampling bag with writing area for easy labeling.
  • Water Sampling Kit
  • Includes sodium thiosulfate tablet
  • Clear, with writing area for easy labeling
Specifications
Color Clear with White Block
Volume (Metric) 300 mL
Includes 10 mg Sodium Thiosulfate Tablet