missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Elgas™ Water Purifier
Click to view available options
Flow Rate:
15 L/hr.
7 L/hr.
Unit Size:
Each
Description
- Purification technologies include pre-treatment, reverse osmosis, and ion exchange.
- Applications include glassware washing/rinsing, feed to ultra pure water systems, reagent make-up/dilution, buffer and media preparation, general chemistry, qualitative analyses and feed to environmental cabinets.
Specifications
Specifications
| Conductivity | 1400μS/cm |
| Flow Rate | 15 L/hr. |
| Type | Water Purifier |
| Temperature Range (Metric) | 1°C to 35°C |
| Width (Metric) | 410 mm |
| Height (Metric) | 460 mm |
| Depth (Metric) | 270 mm |
| Weight (Metric) | 14.5 kg |
| Electrical Requirements | 100/240 V 50/60 Hz |
| Voltage | 100/240 V |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction
