missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water Pollution Standard 3: K, Mg @ 100; Ba, Ca, Mo, Na @ 500 μg/mL in 2% HNO3, VHG™
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Description
Water Pollution Standard 3: K, Mg @ 100; Ba, Ca, Mo, Na @ 500 μg/mL in 2% HNO3
Specifications
Specifications
| Chemical Name or Material | Magnesium/Potassium/Barium/Calcium/Molybdenum/Sodium |
| CAS | 7439-95-4, 9-7-7440, 7440-39-3, 7440-70-2, 7439-98-7, 7440-23-5 |
| Methods | ICP/ICP-MS |
| Analyte or Component Names | Magnesium/Potassium/Barium/Calcium/Molybdenum/Sodium |
| Shipping Condition | Room Temperature |
| Solvent or Matrix | Dilute HNO3 |
| Quantity | 100 mL |
| Concentration or Composition (by Analyte or Components) | Magnesium 100 μg/mL/Potassium 100 μg/mL/Barium 500 μg/mL/Calcium 500 μg/mL/Molybdenum 500 μg/mL/Sodium 500 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction