missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water Pollution Standard 1: Hg @ 5; Cd, Se @ 25; As, Be, Co, Cr, Cu, Fe, Mn, Ni, Pb, Zn @ 100; V @ 250; Al @ 500 μg/mL in 5% HNO3, VHG™
Description
Water Pollution Standard 1: Hg @ 5; Cd, Se @ 25; As, Be, Co, Cr, Cu, Fe, Mn, Ni, Pb, Zn @ 100; V @ 250; Al @ 500 μg/mL in 5% HNO3
Specifications
Specifications
| Chemical Name or Material | Mercury/Cadmium/Selenium/Arsenic/Beryllium/Chromium/Cobalt/Copper/Iron/Lead/Manganese/Nickel/Zinc/Vanadium/Aluminium |
| CAS | 7439-97-6, 7440-43-9, 7782-49-2, 7440-38-2, 7440-41-7, 7440-47-3 |
| Methods | ICP/ICP-MS |
| Analyte or Component Names | Mercury/Cadmium/Selenium/Arsenic/Beryllium/Chromium/Cobalt/Copper/Iron/Lead/Manganese/Nickel/Zinc/Vanadium/Aluminium |
| Shipping Condition | Room Temperature |
| Solvent or Matrix | Dilute HNO3 |
| Quantity | 500 mL |
| Concentration or Composition (by Analyte or Components) | Mercury 5 μg/mL/Cadmium 25 μg/mL/Selenium 25 μg/mL/Arsenic 100 μg/mL/Beryllium 100 μg/mL/Chromium 100 μg/mL/Cobalt 100 μg/mL/Copper 100 μg/mL/Iron 100 μg/mL/Lead 100 μg/mL/Manganese 100 μg/mL/Nickel 100 μg/mL/Zinc 100 μg/mL/Vanadium 250 μg/mL/Aluminium 500 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?