missing translation for 'onlineSavingsMsg'
Learn More

Water Pollution QC Check Sample: Minerals

Product Code. 30327859
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30327859 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30327859 Supplier VHG Labs Supplier No. VHGQWPMIN500
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Alkalinity∣Chloride∣Conductivity∣Fluoride∣Potassium∣Sodium∣Sulfate∣Total dissolved solids∣Total Solids
CAS 16887-00-6, 16984-48-8, 9-7-7440, 7440-23-5, 14808-79-8
Analyte or Component Names Alkalinity∣Chloride∣Conductivity∣Fluoride∣Potassium∣Sodium∣Sulfate∣Total dissolved solids∣Total Solids
Conductivity 393 μmhos/cm
Shipping Condition Room Temperature
Solution Type Mixture
Solvent or Matrix Water
Industry Type Petrochemical Testing
Quantity 500 mL
Concentration or Composition (by Analyte or Components) Fluoride 2.14 mg/L∣Sulfate 12.6 mg/L∣Potassium 14.7 mg/L∣Alkalinity 37.2 mg/L∣Sodium 70.4 mg/L∣Chloride 83.5 mg/L∣Total dissolved solids 248 mg/L∣Total Solids 251 mg/L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.