missing translation for 'onlineSavingsMsg'
Learn More

Water, LC-MS Grade

Product Code. 17875524
Change view
Click to view available options
Quantity:
4 x 4 L
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17875524 4 x 4 L Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17875524 Supplier Thermo Scientific Alfa Aesar Supplier No. 047146.M6
  • ONLINE EXCLUSIVE PRICE
    £277.00
    £238.65 / Pack of 4
    Save £38.35
    13% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used as mobile phase solvent in LC-MS analysis | CAS: 7732-18-5 | H2O | 18.015 g/mol

It is used as mobile phase solvent in LC-MS analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

pH 7 (at 20°C)
Color Colorless
Melting Point 0°C
Boiling Point 100°C
Beilstein 2050024
Solubility Information Soluble in water; Insoluble in polar liquids.
Formula Weight 18.02
Grade LC-MS
Odor Odorless
Density 1 g/cm3
Quantity 4 x 4 L
Refractive Index 1.34
Physical Form Liquid
Chemical Name or Material Water
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.