missing translation for 'onlineSavingsMsg'
Learn More

Water, for Karl Fischer Analysis, Solution In Methanol (1Ml = 5.0mg/H2O), Fisher Chemical™

Product Code. 10763694 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber glass bottle
Plastic Coated Glass bottle
Unit Size:
1 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10763694 1 L Amber glass bottle 1 L
10598050 500 mL Plastic Coated Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10763694 Supplier Fisher Chemical Supplier No. J/9000/15
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

MDL Number 11332

Specifications

Boiling Point 64.7°C
Color Undesignated
Physical Form Liquid
Quantity 1 L
Packaging Amber glass bottle
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Water solution Karl Fischer Laboratory Reagent
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.