Learn More
Specac™ Water Heating Jacket for Liquid and Solid Transmission Sampling
Product Code. 15290238
Encompass_Preferred
Click to view available options
Type:
Water Heating Jacket
Product Code. 15290238
Brand: Specac™ GS20710
This Water Heating Jacket is supplied with a 3in. x 2in. mount to enable the Jacket to be fitted straight into an FTIR instrument. The Water Heating Jacket is similar to the Electrical Heating Jacket, but it uses circulating water to heat the sample cell. A jacket around the circular aperture is filled with a solution, for example water, heated by a thermocirculating system. Temperature control of the sample holder is reliant upon this thermocirculating system. The sample cell holders used with Electrical Heating Jacket can also be used in this accessory.
Specifications
Type Water Heating Jacket